Learn More
Axygen™ Storage Microplates



For sample collection, in-vitro growth chambers and long term storage.
Brand: Axygen™ P-DW-10ML-24-C-S
248.04 GBP valid until 2025-03-31
Use promo code "24073" to get your promotional price.
Description
Used for sample collection, in vitro growth chambers and long-term storage.
Axygen™ deep well and assay plates are used for sample collection, in vitro growth chambers and long-term storage. Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to -80°C.
Specifications
Standard | |
Autoclavable | |
Axygen | |
Microplate | |
10,000 μL | |
Sterile | |
Sample collection, in-vitro growth chambers and long term storage | |
25/Cs. | |
-112°F |
4 x 6 | |
25 Cases | |
Clear | |
Polypropylene | |
24 | |
Square | |
4000 x g | |
5/Pk. | |
-80°C |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
33024067 | Certificate of Quality | 30/03/2025 | P-DW-10ML-24-C-S |
32824041 | Certificate of Quality | 30/03/2025 | P-DW-10ML-24-C-S |
32924017 | Certificate of Quality | 20/03/2025 | P-DW-10ML-24-C-S |
32724055 | Certificate of Quality | 04/03/2025 | P-DW-10ML-24-C-S |
32424129 | Certificate of Quality | 04/03/2025 | P-DW-10ML-24-C-S |
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Days
13Hours
3Minutes
29Seconds
7New Offer Save Up to 92% -
Corning Matrigel
,
Corning Biocoat
,
Corning CoolProduct
,
Corning Media Tech
,
Corning
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24155Expires: 2025-04-30
Days
30Hours
3Minutes
29Seconds
7
Your input is important to us. Please complete this form to provide feedback related to the content on this product.