Learn More
Axygen™ Automation Micropipette Tips
Axygen™ Automation Micropipette Tips are constructed of polypropylene, and feature a variety of available options to suit your needs.
Brand:  Axygen™ ZT-200-R
 View more versions of this product
Product Code. 12033939
  • £308.00 / Pack of 4800
Estimated Shipment: 24-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For Tecan™/Freedom EVO&trade, Te-Mo™, Aquarius, Zymark™/Caliper, RapidPlate™, and SciClone™ Systems
  • For 96-well, Non-ANSI and ANSI/SLAS (384) format, number 5 polypropylene racks
  • Available with or without aerosol-resistant filters
  • Available sterile/non-sterile
  • Available in standard or Maxymum Recovery™ style
Order Info
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
TRUSTED_SUSTAINABILITY
Specifications
Specifications
200 μL
Non-sterile
Blue
RNase-free, DNase-free, Nonpyrogenic
Automation, Round
50 Racks/Cs.
Tecan/Freedom EVO, Te-Mo, Aquarius, Zymark/Caliper, RapidPlate, and SciClone Systems
Non-filtered
Clear
Autoclavable
Rack
96 tips/rack, 10 racks/pack, 5 packs/case
Automation Micropipette Tip
Polypropylene