missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Zymark™ Rapid Plate/Sciclone and Tecan™ TEMO Acquarius Genmate: 50μL
Clear, autoclavable tips; color-coded racks.
Brand: Axygen™ ZTF-50-R-S
This item is not returnable.
View return policy
Description
- High throughput screening
- Volume: 50μL
- Rack Color: Light Green
- Color: Clear
- Designed for use with Zymark™ RapidPlate™, Zymark™ SciClone, Tecan™ Te-MO™, Tecan™ Genmate™ and Tecan™ Aquarius™ automated liquid handling systems
- Certified RNase, DNase and Pyrogen-Free
- Format: 96 well
Order Info
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Specifications
50 μL | |
Sterile | |
Light Green | |
Non-ANSI | |
Rack | |
Tecan Freedom EVO, Tecan TeMo, Tecan Aquarius MCA96 head, Zymark/Caliper RapidPlate, SciClone, SciClone HV96, CAS-4200-1ch. 20-150μL/sec. head | |
Polypropylene |
Filtered | |
Clear | |
Autoclavable | |
Maxymum Recovery™ to further improve accuracy and precision while reducing sample loss | |
Automation, Round | |
Caliper Zymark |