Learn More
Axygen™ AxyMats™ Sealing Mats for 500uL 96 Well Plates
Minimize evaporation in wide range of temperatures in PCR and storage applications
£269.37 - £291.00
Product Code | Brand | Sterility | Pre-slit | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sterility | Pre-slit | Price | Quantity & Availability | ||||
12627215
|
Axygen™
AM-500UL-RD |
Non-sterile | No |
£291.00
Pack of 50 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
||||
12551328
|
Axygen™
AM-500UL-RD-S |
Sterile | No |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Axygen AxyMat sealing mats can be used in a wide range of temperatures for PCR and storage applications. AxyMats minimize evaporation and are made of research-grade silicone which can be autoclaved. They can be reused following a bleach wash and ethanol rinse protocol.
Specifications
Silicone | |
Autoclavable | |
96 |