Learn More
Axygen™ Axypet™ Single Channel Pipettor, Calibration 3x10 STD
High levels of accuracy and precision with design innovations to make pipetting more comfortable during prolonged periods of time.
Brand:  Axygen™
241.45 GBP valid until 2024-12-31
Use promo code "23272" to get your promotional price.
Additional Details : Weight : 0.23080kg
 View more versions of this product
Product Code. 17223624
  • £260.00 / Each
Estimated Shipment: 11-12-2024
to see stock.
Add to basket
Description
Description
  • ISO 17025 Pre-calibrated
  • Thumb-friendly soft spring pipetting system reduces fatigue
  • Unique side EZIject button option reduces thumb movement (Left handed EZIject button available by special order).
  • Finger-hook provides for a more relaxed grip
  • Large ergonomic push button is easy on the thumb during contact
  • Locking feature prevents accidental volume changes during pipetting procedures
Specifications
Specifications
ISO17025
Axypet™
2 to 20 μL
1
Single Channel Pipettor
1/Pk., 1/Cs