Learn More
Axygen™ Axypet™ Starter Kit
CORNING AXYGEN™ Axypet™ Starter Kit includes a set of micropipettes, tips, and other quality accessories.
Brand: Axygen™ AP-STR-KIT-2
470.21 GBP valid until 2024-12-31
Use promo code "23272" to get your promotional price.
Additional Details : Weight : 1.13000kg
Descripción
- Micropipette starter kit
- Includes micropipettes, tips, calibration tools, identification tools, and more
Especificaciones
Axypet | |
10 to 1000 μL | |
1 Kit |
Starter Kit, Micro | |
AP-10, AP-100, AP-1000 micropipettes, 1 rack each of 10μL, 200μL, and 1000μL tips, a calibration tool, ejector spacers, identification labels, lubricant, color identification rings, right-hand ejector button |