missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azadibenzocyclooctyne-NHS ester, with 4 carbon linker, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mg
100 mg
Description
Azadibenzocyclooctyne-NHS ester, with 4 carbon linker is used in various applications like protein-peptide conjugates, antibody-enzyme or antibody-drug conjugates, protein or peptide-oligonucleotide conjugates and surface modification. It is used to introduce cyclooctyne moiety in to biomolecules and amine containing compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Azadibenzocyclooctyne-NHS ester, with 4 carbon linker is used in various applications like protein-peptide conjugates, antibody-enzyme or antibody-drug conjugates, protein or peptide-oligonucleotide conjugates and surface modification. It is used to introduce cyclooctyne moiety in to biomolecules and amine containing compounds.
Solubility
Soluble in dimethyl formamide, terahydrofuran, dimethyl sulfocide and dichloromethane.
Notes
Moisture and heat sensitive. Store in cool place. Incompatible with strong oxidizng agents.
Azadibenzocyclooctyne-NHS ester, with 4 carbon linker is used in various applications like protein-peptide conjugates, antibody-enzyme or antibody-drug conjugates, protein or peptide-oligonucleotide conjugates and surface modification. It is used to introduce cyclooctyne moiety in to biomolecules and amine containing compounds.
Solubility
Soluble in dimethyl formamide, terahydrofuran, dimethyl sulfocide and dichloromethane.
Notes
Moisture and heat sensitive. Store in cool place. Incompatible with strong oxidizng agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | White to Gray |
Quantity | 25 mg |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in dimethyl formamide,terahydrofuran,dimethyl sulfocide and dichloromethane. |
Formula Weight | 402.4 |
Physical Form | Solid |
Chemical Name or Material | Azadibenzocyclooctyne-NHS ester, with 4 carbon linker |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'