missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azadibenzocyclooctyne-PEG, MW 5,000, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mg
Description
Azadibenzocyclooctyne-PEGis used for copper-free click chemistry reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Azadibenzocyclooctyne-PEGis used for copper-free click chemistry reactions.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool,
Azadibenzocyclooctyne-PEGis used for copper-free click chemistry reactions.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool,
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | White to Gray |
Quantity | 100 mg |
Synonym | ADIBO-PEG, MW 5,000 |
Solubility Information | Soluble in water. |
Formula Weight | 5kDa |
Physical Form | Solid |
Chemical Name or Material | Azadibenzocyclooctyne-PEG |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'