missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AZD 6482
GREENER_CHOICE

Product Code. 30001523
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001523

Brand: Tocris Bioscience™ 4839/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective PI 3-Kβ inhibitor

Potent and selective inhibitor of PI 3-kinase β (PI 3-Kβ) (IC50 values are 0.69, 13.6, 47.8 and 136 nM for PI 3-Kβ, PI 3-Kδ, PI 3-Kγ and PI 3-Kα, respectively). Selectively inhibits in vivo growth of the PTEN-deficient tumor xenografts HCC70 and PC3. Also exhibits antithrombotic activity.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1173900-33-8
Molecular Formula C22H24N4O4

Specifications

Quantity 10 mg
Formula Weight 408.45
Purity >98%
Molecular Weight (g/mol) 408.458
Chemical Name or Material AZD 6482
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.