missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AZD 6482, Tocris Bioscience™
missing translation for 'orderingAttributeHoverText'
Quantity:
10 mg
50 mg
Description
Potent and selective inhibitor of PI 3-kinase β (PI 3-Kβ) (IC50 values are 0.69, 13.6, 47.8 and 136 nM for PI 3-Kβ, PI 3-Kδ, PI 3-Kγ and PI 3-Kα, respectively). Selectively inhibits in vivo growth of the PTEN-deficient tumor xenografts HCC70 and PC3. Also exhibits antithrombotic activity.
Specifications
Quantity | 50 mg |
Formula Weight | 408.45 |
Percent Purity | >98% |
Chemical Name or Material | AZD 6482 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'