Learn More
AZD5904, MedChemExpress
MedChemExpress AZD5904 is a selective and irreversible inhibitor of human Myeloperoxidase (MPO) with an IC50 of 140 nM and has similar potency in mouse and rat.

POA - £816.75
Specifications
Chemical Name or Material | AZD5904 |
---|---|
CAS | 618913-30-7 |
Color | Yellow |
Purity Grade Notes | Research |
Molecular Formula | C10H12N4O2S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16484418
![]() |
Medchem Express
HY-111341 |
10 mg |
£228.69
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16450737
![]() |
Medchem Express
HY-111341 |
50 mg |
£816.75
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16423076
![]() |
Medchem Express
HY-111341 |
25 mg |
£456.17
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16417147
![]() |
Medchem Express
HY-111341 |
5 mg |
£168.19
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16405316
![]() |
Medchem Express
HY-111341 |
100 mg |
POA
100mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
AZD5904 is a selective and irreversible inhibitor of human Myeloperoxidase (MPO) with an IC50 of 140 nM and has similar potency in mouse and rat.Specifications
AZD5904 | |
Yellow | |
C10H12N4O2S | |
O=C1NC(N(C[C@@H]2OCCC2)C3=C1N=CN3)=S | |
252.29 | |
Research | |
Neuroscience-Neuromodulation |
618913-30-7 | |
Research | |
DMSO : 22.73 mg/mL (90.09 mM; Need ultrasonic) | |
252.29 | |
98.0% | |
Solid |
Safety and Handling
H361
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.