missing translation for 'onlineSavingsMsg'
Learn More
Azelaic acid, 98%, Thermo Scientific Chemicals Product Code.: 10477022

Azelaic acid, 98%, Thermo Scientific Chemicals

Product Code. 10477022
100 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10477022

Brand: Thermo Scientific Acros 401521000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-99-9
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.223
MDL Number MFCD00004432
InChI Key BDJRBEYXGGNYIS-UHFFFAOYSA-N
Synonym azelaic acid, finacea, anchoic acid, azelex, lepargylic acid, skinoren, 1,7-heptanedicarboxylic acid, heptanedicarboxylic acid, 1,9-nonanedioic acid, emerox 1110Show MoreShow Less
PubChem CID 2266
ChEBI CHEBI:48131
IUPAC Name nonanedioic acid
SMILES C(CCCC(=O)O)CCCC(=O)O

Specifications

Melting Point 98.0°C to 103.0°C
Boiling Point 237.0°C (20.0hPa)
Color White to Yellow
Assay Percent Range 98%
Linear Formula HOOC(CH2)7COOH
Solubility Information Solubility in water: 2.4g/L water (20°C). Other solubilities: very soluble in alcohol and ether:2.7g/100mL(15°C), soluble in organic solvents, slightly soluble in benzene
Formula Weight 188.22
Percent Purity 98%
Quantity 100 g
Flash Point 215°C
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Slightly Crystalline Powder or Flakes
Chemical Name or Material Azelaic acid, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468102 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number401520250,401525000,401521000
Lot NumberA0469022 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number401520250,401525000,401521000
Lot NumberA0464352 Certificate TypeCertificate of Analysis Date18/07/2024 Catalog Number401525000,401520250,401521000
Lot NumberA0463274 Certificate TypeCertificate of Analysis Date30/05/2024 Catalog Number401520250,401525000,401521000
Lot NumberA0424844 Certificate TypeCertificate of Analysis Date04/12/2023 Catalog Number401520250,401525000,401521000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Azelaic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.