missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azelastine hydrochloride, MedChemExpress
Click to view available options
Quantity:
100 mg
200 mg
Unit Size:
100mg
200mg
Description
Azelastine hydrochloridem, an antihistamine, is a potent and selective histamine 1 (H1) antagonist. Azelastine hydrochloride can be used for the research of allergic rhinitis, asthma, diabetic hyperlipidemic and SARS-CoV-2.
Specifications
Specifications
| Chemical Name or Material | Azelastine hydrochloride |
| CAS | 79307-93-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C22H25Cl2N3O |
| Quantity | 200 mg |
| Solubility Information | DMSO : 50 mg/mL (119.51 mM; Need ultrasonic) ∣H2O : 6.67 mg/mL (15.94 mM; Need ultrasonic) |
| SMILES | O=C1N(C2CCN(C)CCC2)N=C(CC3=CC=C(Cl)C=C3)C4=C1C=CC=C4.Cl |
| Molecular Weight (g/mol) | 418.36 |
| Formula Weight | 418.36 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction