missing translation for 'onlineSavingsMsg'
Learn More
Azidotri-n-butyltin(IV), 95%, Thermo Scientific Chemicals Product Code.: 15431948

Azidotri-n-butyltin(IV), 95%, Thermo Scientific Chemicals

Product Code. 15431948
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 15431948

Brand: Thermo Scientific Alfa Aesar H55634.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Azidotri-n-butyltin(IV) is widely utilized as a reagent in the synthesis of tetrazolylbenzene compounds. It is employed in pharmaceutical research and involves synthesis of tetrazole derivatives, which is used to create angiotensin II receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azidotri-n-butyltin(IV) is widely utilized as a reagent in the synthesis of tetrazolylbenzene compounds. It is employed in pharmaceutical research and involves synthesis of tetrazole derivatives, which is used to create angiotensin II receptor antagonists.

Solubility
Miscible with toluene, hexane, acetonitrile and tetrahydrofuran.

Notes
Use in a fume hood. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17846-68-3
Molecular Formula C12H27N3Sn
Molecular Weight (g/mol) 332.079
MDL Number MFCD00216557
InChI Key JKVRTUCVPZTEQZ-UHFFFAOYSA-N
Synonym tributyltin azide, azidotributyltin iv, azidotributyltin, azidotributyl tin, tri-n-butylazidotin, azido tributyl stannane, azido tributyltin, tri-butyltin azide, tributyl tin azide, tributylstannylazide
PubChem CID 4984872
IUPAC Name azido(tributyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)N=[N+]=[N-]

Specifications

Density 1.212
Flash Point 113°C (235°F)
Linear Formula [CH3(CH2)3]3SnN3
Quantity 5 g
UN Number UN2788
Solubility Information Miscible with toluene,hexane,acetonitrile and tetrahydrofuran.
Formula Weight 332.06
Percent Purity 95%
Chemical Name or Material Azidotri-n-butyltin(IV)

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number81202366 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH55634.06,H55634.03,H55634.14
Lot Number5001R07N Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH55634.06,H55634.03,H55634.14
Lot NumberP09D033 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH55634.06,H55634.03,H55634.14
Lot Number5005F10U Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH55634.06,H55634.03,H55634.14
Lot NumberM15G002 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH55634.06,H55634.03,H55634.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Azidotri-n-butyltin(IV), 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.