missing translation for 'onlineSavingsMsg'
Learn More

Azidotrimethylsilane, 94%, Thermo Scientific Chemicals

£57.39 - £749.73

Chemical Identifiers

CAS 4648-54-8
Molecular Formula C3H9N3Si
Molecular Weight (g/mol) 115.21
MDL Number MFCD00001986
InChI Key SEDZOYHHAIAQIW-UHFFFAOYSA-N
Synonym trimethylsilyl azide, silane, azidotrimethyl, trimethylsilylazide, azido trimethyl silane, ccris 8048, tms azide, tms-azide, trimethyl silylazide, trimethylsilyl-azide, azido trimethylsilane
PubChem CID 78378
IUPAC Name azido(trimethyl)silane
SMILES C[Si](C)(C)N=[N+]=[N-]
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10429270
View Documents Promotion Details
Thermo Scientific Acros
167930100
10 mL Glass bottle
£57.39
10mL
Please to purchase this item. Need a web account? Register with us today!
10763551
View Documents
Thermo Scientific Acros
167930500
50 mL Glass bottle
£191.96
50mL
Please to purchase this item. Need a web account? Register with us today!
11985801
View Documents
Thermo Scientific Acros
167932500
250 mL Glass bottle
£749.73
250mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, For Azidation/Diazo transfer
Specifications

Chemical Identifiers

4648-54-8
115.21
SEDZOYHHAIAQIW-UHFFFAOYSA-N
78378
C[Si](C)(C)N=[N+]=[N-]
C3H9N3Si
MFCD00001986
trimethylsilyl azide, silane, azidotrimethyl, trimethylsilylazide, azido trimethyl silane, ccris 8048, tms azide, tms-azide, trimethyl silylazide, trimethylsilyl-azide, azido trimethylsilane
azido(trimethyl)silane

Specifications

4648-54-8
0.8760g/mL
6°C
92% min. (GC)
(CH3)3SiN3
MFCD00001986
0.876
Solubility in water: decomposes. Other solubilities: soluble in ether, dichloromethane and benzene
C[Si](C)(C)N=[N+]=[N-]
115.21
115.21
Liquid
Colorless to Yellow
95.0°C to 96.0°C
Authentic
C3H9N3Si
1.4142 to 1.4162
01,1236; 03,316; 05,719; 06,632; 07,394; 09,21; 10,14; 11,32; 12,37; 13,24; 14,25; 15,16; 16,17; 17,378
trimethylsilyl azide, silane, azidotrimethyl, trimethylsilylazide, azido trimethyl silane, ccris 8048, tms azide, tms-azide, trimethyl silylazide, trimethylsilyl-azide, azido trimethylsilane
SEDZOYHHAIAQIW-UHFFFAOYSA-N
azido(trimethyl)silane
78378
94%
Azidotrimethylsilane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
Toxic in contact with skin.
Toxic if inhaled.
Highly flammable liquid and vapor.
Contact with water liberates toxic gas.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 225-078-5

Product Identifier
  • Azidotrimethylsilane

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Flammable liquid Category 2

Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H225-Highly flammable liquid and vapour.
  • EUH029-Contact with water liberates toxic gas.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.