Learn More
Azlon™ Gas Delivery Tube
Polypropylene construction. Azlon™ Gas Delivery Tube are ideal substitutes for fragile glass alternatives.
£33.85 - £66.75
Specifications
Material Polypropylene
For Use With (Application) Many types of gas collection experiments in schools and universities
Max. Operating Temperature (Metric) 135°C
View More Specs
Products 4
Product Code Brand Overall Length (Metric) Product Type Price Quantity & Availability  
Product Code Brand Overall Length (Metric) Product Type Price Quantity & Availability  
12673012
View Documents
Azlon™
DT002
290 mm Bung Top Gas Delivery Tube This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12683012
View Documents
Azlon™
DT003
335 mm Side Arm Gas Delivery Tube This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12663012
View Documents
Azlon™
DT001
205 mm Gas Delivery Tube This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12693012
missing translation for 'viewDocuments'
Azlon™
DT005
475 mm Double Bend Gas Delivery Tube This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Pre-bent rigid polypropylene delivery tubes
  • Unbreakable in normal use
  • Suitable for many types of gas collection experiments in schools and universities
Spécification
Spécification
Polypropylene
Many types of gas collection experiments in schools and universities
135°C
Vidéos
FDS
Documents
Documents
Certificats
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Azlon™ Gas Delivery Tube
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis