missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlon™ LDPE Gram Stain Wash Bottles
36.02 GBP valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Use promo code "23259" to get your promotional price.
Click to view available options
Capacity (Metric):
250 mL
Description
Pre-labelled with Gram stain solutions, hazard warning labels and instructions for use
- Pre-labelled with Gram stain solutions, hazard warning labels and instructions for use
- With patented DripLok™ vapour venting technology
- The set of five wash bottles are printed with: - Crystal violet solution - Lugol's iodine solution - Gram iodine solution - Gram color/de-colorising solution - Safranine solution
Specifications
Capacity (Metric) | 250 mL |
Capacity (English) | 8.45 oz. |
Color | Translucent |
For Use With (Application) | For Gram staining |
Includes | Five wash bottles printed with the following information: crystal violet solution, Lugol's Iodine solution, Iodine solution Gram, Gram-color/de-colorizing solution, Safranin solution |
Material | LDPE |
Temperature Range (Metric) | –50 to 80°C |
Closure Included | Yes |
Autoclavable | Not Autoclavable |
Graduated | No |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'