missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ LDPE Printed Slope Shoulder Wash Bottles

Product Code. p-8001804
Click to view available options
Capacity (Metric):
250 mL
500 mL
Chemical Label:
Acetone
Distilled Water
Eau distillée
Ethanol
I. M. S.
Isopropanol
Methanol
Water
Closure Color:
Blue
Green
Orange
Red
White
Yellow
Unit Size:
Each
Pack of 5
This item is not returnable. View return policy
Wash bottles in this range that are prelabelled for intended usage with specific hazardous substances are also prelabelled with the appropriate GHS hazard warning symbols.
 

Product Code. 11353514

Brand: Azlon™ WGF535P

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Wash bottles in this range that are prelabelled for intended usage with specific hazardous substances are also prelabelled with the appropriate GHS hazard warning symbols.
 

Use these durable plastic bottles for a wide range of liquid storage, media preparation and sampling applications. Azlon™ LDPE Printed Slope Shoulder Wash Bottles are indelibly printed with a choice of seven popular laboratory solvents, appropriate hazard symbols and CAS numbers.

Use these durable plastic bottles for a wide range of liquid storage, media preparation and sampling applications. AzlonTM LDPE Printed Slope Shoulder Wash Bottles are indelibly printed with a choice of seven popular laboratory solvents, appropriate hazard symbols and CAS numbers.

  • Labeled wash bottles for easy identification of contents
  • Slim design enables comfortable handling and enhanced ease of use
  • Fitted with color-coded polypropylene closures
  • Closures feature precision jet-tip aperture for fine control
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 250 mL
Capacity (English) 8.45 oz.
Color Translucent
Diameter (Metric) Outer Neck 54mm
For Use With (Application) For easy identification of contents
Includes Orange Polypropylene closure
Material LDPE
No. per Pack 5/PK
Closure Color Orange
Closure Included Yes
Graduated No
Mouth Narrow
Shape Round
Height (Metric) 152 mm
Product Type Wash Bottle
Chemical Label Ethanol
Closure Material Polypropylene
Diameter (Metric) 152 mm
Closure Type Cap
Through the Closure Dispensing Yes
Autoclavable Not Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.