missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlon™ Lightweight Polypropylene Funnels
Lightweight polypropylene made from food grade plastic
Brand: Azlon FWC062
41.68 GBP valid until 2024-11-29
Use promo code "21718" to get your promotional price.
This item is not returnable.
View return policy
Description
Manufactured from lightweight polypropylene. Azlon™ Lightweight Polypropylene Funnels include ribs that prevent airlocks and allow rapid filtration. Autoclavable to 121 °C.
- Lightweight polypropylene funnels with ribs that prevent air locks, allowing rapid filtration
- Autoclavable to 121°C
- Made from food grade plastic
Specifications
13mm | |
Polypropylene | |
149 mm | |
Autoclavable |