Learn More
Azlon™ Magnetic Retrievers for Stirrer Bars
Retrieve stirrer bars from sensitive liquids with these magnetic retrievers. Azlon™ Magnetic Retrievers for Stirrer Bars feature magnets permanently embedded in a PTFE rod.
£44.90 - £52.40
Specifications
Autoclavable Autoclavable
Coating PTFE
Diameter (Metric) 10 mm
Material Magnet
Quantity 1/Pk.
View More Specs
Products 2
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
10514355
View Documents
Azlon™
SWN564
250 mm
£44.90
Each
In Stock
to see stock.
Edge
Add to basket
10400095
View Documents Promotion Details
Azlon™
SWN566
350 mm
£52.40
Each
In Stock
to see stock.
Edge
Add to basket
Description
Description
Retrieve stirrer bars from sensitive liquids with these magnetic retrievers. AzlonTM Magnetic Retrievers for Stirrer Bars feature magnets permanently embedded in a PTFE rod.
  • Ideal for retrieving magnetic stirrer bars from particularly sensitive or corrosive liquids
  • Turbo retrievers are ideal for use with turbo stirrer bars
Specifications
Specifications
Autoclavable
10 mm
1/Pk.
PTFE
Magnet