missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlon™ Micro Magnetic Stirrer Bar
Use these bars in for a variety of stirring applications. Fisherbrand™ Azlon™ Micro Magnetic Stirrer Bars are ideal for small vessels, test tubes and vials.
Brand: Azlon SWN956
8.75 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.
Description
Use these bars in for a variety of stirring applications. Fisherbrand™ Azlon™ Micro Magnetic Stirrer Bars are ideal for small vessels, test tubes and vials.
- For efficient stirring
- PTFE composition
Specifications
Autoclavable | |
3 mm | |
Magnet | |
Micro |
0.51 in. | |
13 mm | |
2/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction