missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Oval Magnetic Stirrer Bar

Use these bars in for a variety of stirring applications. Azlon™ Oval Magnetic Stirrer Bars are ideal for round bottomed flasks.

Brand:  Azlon SWN525

44.32 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10779345

  • £67.65 / Pack of 2
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For efficient stirring
  • PTFE composition
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
16 mm
Magnet
Oval
1.3 in.
35 mm
2/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.