missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Square Polycarbonate Bottles

Product Code. 15364941
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
2000 mL
Diameter (Metric) Inner Neck:
28 mm
40 mm
missing translation for 'unitSize'
Pack of 12
Pack of 24
Pack of 6
This item is not returnable. View return policy

Product Code. 15364941

missing translation for 'mfr': Azlon BSC0250

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Ideal for preparation and storage of biological media, reagents and aqueous solutions

  • Highly transparent polycarbonate media bottles with linerless polypropylene cap
  • Suitable for repeated sterilisation by autoclaving at 121°C
  • Leakproof
  • Practical square design for economic use of space
  • Heavy duty and durable in use
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 250 mL
Diameter (Metric) Inner Neck 28 mm
Material Polycarbonate
Neck ID (Metric) 28mm
Quantity 24/Pk.
Shape Square
Height (Metric) 139 mm
Product Type Square Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.