Learn More
Azlon™ Polyethylene Stoppers
Use these stoppers with glass or plastic volumetric flasks and measuring cylinders which have longer 'B' length sockets. Azlon™ Polyethylene Stoppers feature a large head for a good grip and extra protection if glassware is knocked over.
Brand:  Azlon™ TRF593
19.39 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11317804
  • £44.30 / Pack of 10
Estimated Shipment: 09-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Use these stoppers with glass or plastic volumetric flasks and measuring cylinders which have longer 'B' length sockets. AzlonTM Polyethylene Stoppers feature a large head for a good grip and extra protection if glassware is knocked over.
  • Manufactured from polyethylene
  • Available in white and red
  • Suitable for use with both Azlon and glass volumetric ware
  • Enlarged head provides a good grip and protects glassware
  • Ribbed cone ensures a secure leak free seal
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Glass or Plastic volumetric flasks and measuring cylinders etc
White, Red
Tapered Stopper
14/23
Polyethylene
Ribbed Cone
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Azlon™ Polyethylene Stoppers > Cone Size: 14/23; Joint Size: 14/23