Learn More
Azlon™ Polypropylene Measuring Scoop
Scoop chemical reagents or food items safely. The Azlon™ polypropylene measuring scoop comes with flat top and bottom.
£3.60 - £90.65
Product Code | Brand | Capacity (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
11349633
|
Azlon
SWL140 |
1000 mL | 385 mm |
£31.15
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
12511516
|
Azlon
SWL008 |
10 mL | 102 mm |
£9.70
Pack of 5 |
Estimated Shipment: 25-09-2024
Log in to see stock. |
||||
11333724
|
Azlon
SWL024 |
100 mL | 198 mm |
£68.01
Pack of 5 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
12521516
|
Azlon
SWL030 |
250 mL | 259 mm |
£72.81
Pack of 5 |
In Stock
Log in to see stock. |
||||
11313724
|
Azlon
SWL012 |
25 mL | 137 mm |
£33.75
Pack of 5 |
Estimated Shipment: 25-09-2024
Log in to see stock. |
||||
11359623
|
Azlon
SWL036 |
500 mL | 315 mm |
£90.65
Pack of 5 |
Estimated Shipment: 06-08-2024
Log in to see stock. |
||||
11323724
|
Azlon
SWL018 |
50 mL | 160 mm |
£41.60
Pack of 5 |
In Stock
Log in to see stock. |
||||
11357784
|
Azlon
SWL030 |
250 mL | 259 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12927325
|
Azlon
SWL036 |
500 mL | 315 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12591506
|
Azlon
SWL002 |
2 mL | 60 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
12501516
|
Azlon
SWL006 |
5 mL | 82 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Scoop chemical reagents or food items safely. The Azlon™ polypropylene measuring scoop comes with flat top and bottom.
- Made from food safe polypropylene
- Top is flat for levelling off powders
- Base is flat enabling scoops to stand horizontally
- The capacity is clearly molded on each size
Specifications
Measuring Scoop | |
Polypropylene | |
Handle |