missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Polypropylene One Mark Class B Pipets

Measure liquids with precision. The Azlon™ Polypropylene One Mark Class B pipets are designed for accuracy.

£29.00 - £307.00

Specifications

Calibration Calibrated to Class B tolerances
Class B
Material Polypropylene
Graduated Yes
Color Translucent
View More Specs

Products 6
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
11363704
View Documents
Azlon
PDH610
10 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11373704
View Documents
Azlon
PDH625
25 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11383704
View Documents
Azlon
PDH650
50 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10399794
View Documents
Azlon
PDH601
1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10440865
View Documents
Azlon
PDH605
5 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10208024
View Documents
Azlon
PDH602
2 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Measure liquids with precision. The Azlon™ Polypropylene One Mark Class B pipets are designed for accuracy.

  • Calibrated to meet Class B tolerances
  • Permanently marked with ring and inscriptions
Specifications

Specifications

Calibrated to Class B tolerances
Polypropylene
Translucent
B
Yes
Graduated Pipet
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Azlon™ Polypropylene One Mark Class B Pipets

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.