missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Polypropylene Powder Funnels

Constructed of durable, lightweight Polypropylene. Azlon™ Polypropylene Powder Funnel features a parallel stem that minimises 'bridging' of powder to improve flow. The two largest sizes have vertical-sided tops to prevent spillage.

Brand:  Azlon FWC210

Code : 02

Additional Details : Weight : 0.23688kg

 View more versions of this product

Product Code. 10589730

  • £59.95 / Pack of 5
Estimated Shipment: 20-08-2024
to see stock.
Add to basket

Description

Description

Constructed of durable, lightweight Polypropylene. Azlon™ Polypropylene Powder Funnel features a parallel stem that minimises 'bridging' of powder to improve flow. The two largest sizes have vertical-sided tops to prevent spillage.

  • Lightweight polypropylene powder funnel
  • Unbreakable in normal use
  • Parallel stem minimises 'bridging' of powder to improve flow
  • The two largest sizes have vertical-sided tops
Specifications

Specifications

150mm
Polypropylene
Yes
36mm
135 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers