missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Polypropylene Spatula

Manufactured from polypropylene. SciLabWare™ Azlon™ Polypropylene Spatula is virtually unbreakable in normal use and retain shape after flexing.

Brand:  Azlon SWY062P

 View more versions of this product

Product Code. 11397784

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Manufactured from polypropylene. SciLabWare™ Azlon™ Polypropylene Spatula is virtually unbreakable in normal use and retain shape after flexing.

  • Molded strong tapered spatulas with chamfered ends
  • Virtually unbreakable in normal use and retain shape after flexing
  • Good chemical and heat resistance
  • Autoclavable at 121°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Autoclavable
26 mm
Polypropylene
Spatula
165 x 26 mm
165 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers