missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlon™ Screw Cap for Heavy Duty Carboys
Click to view available options
Color:
White
Description
- Spare caps for heavy duty polypropylene aspirators
Specifications
Color | White |
Quantity | 10/Pk. |
For Use With (Equipment) | Heavy duty polypropylene aspirators |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'