Learn More
Pyrex™ Quickfit™ Stopper for Iodine Flask
For use with Iodine Stopper Flask
Brand: Quickfit SIB29
343.66 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
For use with Iodine Stopper Flask
- Stopper has extended handle which projects well above liquid sea
Specifications
Glass | |
Iodine Flask | |
Stopper | |
29/32 |