missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Tapered Glass Joint Sleeve

Product Code. p-8002921
Click to view available options
Standard Taper Joint No.:
10/19
14/23
19/26
24/29
29/32
34/35
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12063354

Brand: Azlon SWN804

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Forms a liquid and gas tight seal.

  • For grease free and high vacuum seals using Quickfit™ ground glass joints
  • Strong construction with knurled shoulder for ease of handling and repeated use
  • Self lubricating and non-contaminating
  • Working temperature range -100°C to +260°C
  • Working vacuum ≤10-3 atmospheres
  • Highly resistant to chemical attack
TRUSTED_SUSTAINABILITY

Specifications

Material PTFE
Product Type Tapered sleeves
Temperature Range (Metric) –100 to 250°C
Standard Taper Joint No. 14/23
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.