Learn More
Azlon™ USA OSHA Vented Wide Mouth Wash Bottles
Catalog Number 15521232
38.52 GBP valid until 2024-12-31
Use promo code "23259" to get your promotional price.
Click to view available options
Chemical Label:
Acetone
Distilled Water
Ethanol
Isopropanol
Methanol
Multi-lingual wide neck, OSHA, wash bottles featuring the patented DripLok™ vapor venting valve that prevents solvent drips.
  • Indelibly printed in 3 languages (English, French and Spanish)
  • Choose from five solvent labels all including the GHS, NFPA diamond, PPE pictograms, risk and safety information
  • Fitted with color coded one-piece PP closures that maintain their shape in use
  • Closures feature precision jet-tip aperture for fine control
Capacity (Metric) 500 mL
Material Plastic
Quantity 5/Pk.
Chemical Label Acetone
Closure Type Vapor-venting