missing translation for 'onlineSavingsMsg'
Learn More
Azobenzene, 98%, Thermo Scientific Chemicals Product Code.: 10167510

Azobenzene, 98%, Thermo Scientific Chemicals

Product Code. 10167510
50 g, Glass bottle
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10167510

Brand: Thermo Scientific Acros 401540500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-33-3
Molecular Formula C12H10N2
Molecular Weight (g/mol) 182.23
MDL Number MFCD00003022
InChI Key DMLAVOWQYNRWNQ-UHFFFAOYSA-N
Synonym azobenzene, diazene, diphenyl, diazobenzene, e-diphenyldiazene, azobisbenzene, azofume, 1,2-diphenyldiazene, azodibenzeneazofume, diphenyldiimide, azobenzideShow MoreShow Less
PubChem CID 2272
ChEBI CHEBI:190358
IUPAC Name diphenyldiazene
SMILES C1=CC=C(C=C1)N=NC1=CC=CC=C1

Specifications

Melting Point 66.0°C to 69.0°C
Color Orange
Boiling Point 293.0°C
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5N=NC6H5
Quantity 50 g
Beilstein 16, 8
Merck Index 15, 909
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether, glacial acetic acid,, acetone and benzene
Formula Weight 182.23
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Azobenzene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0437415 Certificate TypeCertificate of Analysis Date15/07/2024 Catalog Number401542500,401540500
Lot NumberA0430442 Certificate TypeCertificate of Analysis Date11/12/2023 Catalog Number401542500,401540500
Lot NumberA0449084 Certificate TypeCertificate of Analysis Date11/01/2023 Catalog Number401542500,401540500
Lot NumberA0408518 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number401542500,401540500
Lot NumberA0414662 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number401542500,401540500

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Azobenzene, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.