Learn More
Azomethine-H hydrate, Thermo Scientific Chemicals
Reagent for boron determination
£49.10 - £369.00
Chemical Identifiers
CAS | 32266-60-7 |
---|---|
Molecular Formula | C17H13NO8S2 |
Molecular Weight (g/mol) | 423.41 |
MDL Number | MFCD00066536 |
InChI Key | DUCCKQSNXPFEGT-NVMNQCDNSA-N |
PubChem CID | 5702781 |
SMILES | OC1=CC=CC=C1\C=N/C1=C2C(O)=CC(=CC2=CC(=C1)S(O)(=O)=O)S(O)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11327228
|
Thermo Scientific Alfa Aesar
016810.04 |
2 g |
£49.10
2g |
In Stock
Log in to see stock. |
|||||
11317228
|
Thermo Scientific Alfa Aesar
016810.09 |
10 g |
£182.00
10g |
Estimated Shipment: 17-10-2024
Log in to see stock. |
|||||
11337228
|
Thermo Scientific Alfa Aesar
016810.14 |
25 g |
£369.00
25g |
In Stock
Log in to see stock. |
|||||
Description
Azomethine-H hydrate is used as a highly sensitive and colorimetric reagent for testing and detection of boron. It is also used to detect microgram levels of boron in glass and steel samples. Further, it is involved in the detection of boron in plant samples by masking copper, iron and aluminum ions with ethylenediaminetetraacetic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
32266-60-7 | |
423.41 | |
DUCCKQSNXPFEGT-NVMNQCDNSA-N | |
OC1=CC=CC=C1\C=N/C1=C2C(O)=CC(=CC2=CC(=C1)S(O)(=O)=O)S(O)(=O)=O |
C17H13NO8S2 | |
MFCD00066536 | |
5702781 |
Specifications
32266-60-7 | |
MFCD00066536 | |
DUCCKQSNXPFEGT-NVMNQCDNSA-N | |
4-hydroxy-5-[[(Z)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]naphthalene-2,7-disulfonic acid | |
5702781 | |
Light sensitive | |
Azomethine-H hydrate |
C17H13NO8S2 | |
Soluble in acetone,ammonium hydroxide,n-butanol and water. | |
OC1=CC=CC=C1\C=N/C1=C2C(O)=CC(=CC2=CC(=C1)S(O)(=O)=O)S(O)(=O)=O | |
423.41 | |
423.40 (Anhydrous) | |
Powder |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 250-975-3
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only