Learn More
Azomethine-H monosodium salt hydrate, Thermo Scientific Chemicals
£20.70 - £124.62
Chemical Identifiers
CAS | 206752-32-1 |
---|---|
Molecular Formula | C17H12NNaO8S2 |
Molecular Weight (g/mol) | 445.39 |
MDL Number | MFCD00010498 |
InChI Key | GRYBZNLNHVSHPY-UHFFFAOYSA-M |
Synonym | sodium 5-hydroxy-4-2-hydroxybenzylidene amino-7-sulfonaphthalene-2-sulfonate hydrate, sodium 5-hydroxy-4-e-2-hydroxyphenyl methylidene amino-7-sulfonaphthalene-2-sulfonate hydrate, azomethin-h monosodium salt hydrate, 4-hydroxy-5-e-2-hydroxyphenyl methylidene amino-7-sodiooxysulfonyl naphthalene-2-sulfonic acid hydrate, sodium 5-hydroxy-4-2-hydroxyphenyl methylidene amino-7-sulfonaphthalene-2-sulfonate hydrate |
PubChem CID | 122198137 |
SMILES | [Na+].OC1=C2C(NC=C3C=CC=CC3=O)=CC(=CC2=CC(=C1)S([O-])(=O)=O)S(O)(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11464323
|
Thermo Scientific Alfa Aesar
A10927.03 |
1 g |
£20.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11474323
|
Thermo Scientific Alfa Aesar
A10927.06 |
5 g |
£39.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11484323
|
Thermo Scientific Alfa Aesar
A10927.14 |
25 g |
£124.62
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Azomethine-H monosodium salt hydrate is a reagent used for the colorimetric determination of boron in soil, plants, composts, manure, water and nutrient solutions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAzomethine-H monosodium salt hydrate is a reagent used for the colorimetric determination of boron in soil, plants, composts, manure, water and nutrient solutions.
Solubility
Soluble in water, ethanol and acetone.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at room temperature.
Chemical Identifiers
206752-32-1 | |
445.39 | |
GRYBZNLNHVSHPY-UHFFFAOYSA-M | |
122198137 |
C17H12NNaO8S2 | |
MFCD00010498 | |
sodium 5-hydroxy-4-2-hydroxybenzylidene amino-7-sulfonaphthalene-2-sulfonate hydrate, sodium 5-hydroxy-4-e-2-hydroxyphenyl methylidene amino-7-sulfonaphthalene-2-sulfonate hydrate, azomethin-h monosodium salt hydrate, 4-hydroxy-5-e-2-hydroxyphenyl methylidene amino-7-sodiooxysulfonyl naphthalene-2-sulfonic acid hydrate, sodium 5-hydroxy-4-2-hydroxyphenyl methylidene amino-7-sulfonaphthalene-2-sulfonate hydrate | |
[Na+].OC1=C2C(NC=C3C=CC=CC3=O)=CC(=CC2=CC(=C1)S([O-])(=O)=O)S(O)(=O)=O |
Specifications
206752-32-1 | |
C17H12NNaO8S2 | |
4100175 | |
Soluble in water,ethanol and acetone. | |
[Na+].OC1=C2C(NC=C3C=CC=CC3=O)=CC(=CC2=CC(=C1)S([O-])(=O)=O)S(O)(=O)=O | |
445.39 | |
445.41 (Anhydrous) |
>300°C | |
MFCD00010498 | |
sodium 5-hydroxy-4-2-hydroxybenzylidene amino-7-sulfonaphthalene-2-sulfonate hydrate, sodium 5-hydroxy-4-e-2-hydroxyphenyl methylidene amino-7-sulfonaphthalene-2-sulfonate hydrate, azomethin-h monosodium salt hydrate, 4-hydroxy-5-e-2-hydroxyphenyl methylidene amino-7-sodiooxysulfonyl naphthalene-2-sulfonic acid hydrate, sodium 5-hydroxy-4-2-hydroxyphenyl methylidene amino-7-sulfonaphthalene-2-sulfonate hydrate | |
GRYBZNLNHVSHPY-UHFFFAOYSA-M | |
sodium 4-hydroxy-5-{[(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino}-7-sulfonaphthalene-2-sulfonate hydrate | |
122198137 | |
Azomethine-H monosodium salt hydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 250-975-3
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only