missing translation for 'onlineSavingsMsg'
Learn More

Azomethine H, MP Biomedicals™

Product Code. 11437532
Click to view available options
Quantity:
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 5941-07-1
Molecular Formula C17H14NNaO9S2
Molecular Weight (g/mol) 463.41
MDL Number MFCD00149588
InChI Key BEDIAJJRAYEWBI-TTWKNDKESA-M
Synonym 8-Hydroxy-1-(salicylideneamino)naphthalene-3, 6 disulfonic acid monosodium salt, 4-Hydroxy-5-(2-hydroxybenzylideneamino)-naphthalene-2, 7-disulfonic acid monosodium salt hydrate

Specifications

Assay Percent Range 1.04
Extinction Coefficient Extinction Coefficient max: 237nm C=0.01% ≥50, 000 (Lit.)
SMILES C1=CC(=CNC2=C3C(=CC(=C2)S(=O)(=O)O)C=C(C=C3O)S(=O)(=O)O)C(=O)C=C1.[Na].[Na]
IUPAC Name 4-hydroxy-5-[[(Z)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]naphthalene-2,7-disulfonic acid;sodium
PubChem CID 123134089
Formula Weight 423.4
Color Orange
Quantity 10 g
Physical Form Powder
Chemical Name or Material Azomethine H, Chemistry
compliance-icons
Product Identifier
  • Azomethine H Monosodium Salt Monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Azomethine H Monosodium Salt Monohydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'