missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azpack™ Cargille™ Immersion Oil
Choose between three types for normal or fluorescence microscopy. Azpack™ Cargille™ Immersion Oil is supplied with a listing of optical and physical properties.
£104.00 - £263.00
Code produit | Marque | Type | Viscosity | Quantity | Prix | Quantité et disponibilité | |||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Type | Viscosity | Quantity | Prix | Quantité et disponibilité | |||
12423170
|
Azpack™
12423170 |
Immersion Oil Type B | 1250 cSt | 30 mL |
£104.00
Each |
In Stock
Log in to see stock. |
|||
11338113
|
Azpack™
11338113 |
Immersion Oil Type B | 1250 cSt | 120 mL |
£169.00
Each |
In Stock
Log in to see stock. |
|||
11328113
|
Azpack™
11328113 |
Immersion Oil Type A | 150 cSt | 120 mL |
£160.00
Each |
Estimated Shipment: 14-08-2024
Log in to see stock. |
|||
11358113
|
Azpack™
11358113 |
Immersion Oil Type FF | 170 cSt | 120 mL |
£263.00
Each |
In Stock
Log in to see stock. |
|||
Description
- Supplied with specific optical and physical properties listing
- For use with normal light or fluorescence microscopy
Specifications
Normal light or fluorescence microscopy |