missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azpack™ Quartz Wool
Filter acids and hot gases or use as a stopper for combustion tubes. Azpack™ Quartz Wool can be exposed to temperatures up to 1050°C.
Brand: Azpack™ 12373866
Alert:
Adding fire retardant materials will not stop fibres from fusing together if heated past 1400°C
Description
- A mass of fibres, average diameter 6μm.
- Used for filtering acids and hot gases or as a plug for combustion tubes.
Specifications
1Each | |
Filtering acids, hot gases | |
10g | |
6μm per fiber |
Wool Filter | |
Quartz fiber | |
400°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction