missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ B-27™ Supplement (50X), minus vitamin A

Product Code. 11500446
Click to view available options
Quantity:
10 mL
100 mL
Unit Size:
100mL
10mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11500446

Brand: Gibco™ 12587010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

A variant of the original serum-free B-27 Supplement, designed for the proliferation of neural stem cells in the absence of vitamin A.

Gibco B-27 Supplement minus vitamin A is a customized, serum-free formulation based on the original, most cited neuronal cell culture supplement, designed specifically to support the proliferation and cultivation of neural stem cells (NSCs) and neural progenitor cells. By excluding vitamin A (also known as retinyl acetate or retinol), this variant prevents the differentiation-inducing effects of retinoic acid, making it ideal for maintaining NSCs and neural progenitors in their undifferentiated state.

  • Optimized for NSC proliferation—by removing vitamin A, this supplement is perfectly suited for the growth and expansion of embryonic and adult neural stem cells and neural precursors, without inducing differentiation
  • Versatile cultivation—supports the cultivation of neural progenitor cells and NSCs in both neurospheres in suspension and adherent monolayer cultures, providing flexibility in your experimental design. Ideal for cell culture where retinoic acid signaling must be controlled.
  • High quality—cGMP-manufactured to ensure the highest standards of quality and consistency. Each batch undergoes rigorous performance testing, including assessments of its ability to support the differentiation of human neural stem cells to neurons.
  • Comprehensive testing—every lot is tested for endotoxins, sterility, and mycoplasma to ensure the utmost safety and reliability
  • Proven performance—derived from the proprietary formulation of the original B-27 Supplement, trusted by researchers worldwide for its consistent and reliable results

Choose Gibco B-27 Supplement, minus vitamin A, for an optimized, high-quality solution that supports the proliferation of neural stem cells while preventing unwanted differentiation influenced by retinoic acid, ensuring consistent and reliable results for your research.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -20°C) and protect from light.
Cell Type Neural Progenitor Cells
Classification Serum-free
Concentration 50 X
Culture Type Mammalian Cell Culture
Form Liquid
Product Type Cell Culture Supplement
Serum Level Serum-Free
Sterility Sterile
Product Line B-27
Quantity 10 mL
Shipping Condition Dry Ice
Source Serum-Free
Tested For Endotoxin, Performance, Sterility, Mycoplasma
Show More Show Less
compliance-icons
Product Identifier
  • 5-Methoxyindole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Indol-5-yl methyl ether

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.