missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ B-27™ Supplement (50X), minus vitamin A
GREENER_CHOICE

Suitable for use in most neuronal cell applications

Brand:  Gibco™ 12587001

883.64 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.


 View more versions of this product

Product Code. 15440584

  • £1056.00 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

A variation of the most cited neuronal cell culture supplement, B-27 Supplement Minus Vitamin A is a customized version of the original B-27 Supplement without vitamin A (retinyl acetate).  B-27 Supplement Minus Vitamin A is a serum-free supplement ideal for the cultivation of neural progenitor and stem cells, either as neurospheres in suspension or in adherent monolayer culture, without inducing differentiation.  B-27 Supplement Minus Vitamin A is provided as a 50X liquid and is intended for use with Neurobasal Medium or Neurobasal-A Medium for neuronal cell culture without the need for an astrocyte feeder layer.
 
 
 
B-27 Supplement is suitable for use in most neuronal cell applications. For other applications, we provide alternative formulations of B-27 Supplement and Neurobasal media
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in freezer (-5 to -20°C) and protect from light.
Serum-free
Mammalian Cell Culture
Cell Culture Supplement
Sterile
100 mL
Serum-Free
Neural Progenitor Cells
50 X
Liquid
Serum-Free
B-27™
Dry Ice
Endotoxin, Performance, Sterility
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.