missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ B-27™ Supplement (50X), serum free

Product Code. 11530536
Click to view available options
Quantity:
10 mL
100 mL
Unit Size:
100mL
10mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11530536

Brand: Gibco™ 17504044

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

The original Gibco B-27 Supplement, optimized for serum-free culture of neurons, supports the growth and viability of neurons and many different cell types.

This original and genuine B-27 Supplement is a cGMP-manufactured, serum-free solution designed to ensure the consistent, reliable, and high-quality growth of embryonic, post-natal, and adult hippocampal neurons, as well as other CNS neurons. With a proven track record, B-27 Supplement supports both low- and high-density cultures, ensuring viability in serum-free media. Each batch undergoes rigorous performance testing to confirm its ability to successfully culture hippocampal neurons, providing you with the confidence and dependability you need for your research.

  • Dependable consistency—every lot is performance-tested for reliable results
  • High quality—cGMP manufacturing provides the highest standards of quality
  • Versatile usage—ideal for use with serum-free media such as Neurobasal Medium or Neurobasal-A Medium, without the need for an astrocyte feeder layer. Supports the growth and viability of neurons, as well as many other cell types differentiated from PSCs.
  • Proven performance—a proprietary formulation; the first and most cited neuronal cell culture supplement, trusted by researchers worldwide

Choose B-27 Supplement for the authentic solution that sets the standard for neuronal cell culture.

Order Info

Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5 to -20°C) and protect from light.
Cell Type Neural Cells
Classification Serum-free
Concentration 50 X
Culture Type Mammalian Cell Culture
Form Liquid
Product Type Cell Culture Supplement
Serum Level Serum-Free
Sterility Sterile
pH 6 to 8
Product Line B-27
Quantity 10 mL
Shipping Condition Dry Ice
Source Serum-Free
Tested For Endotoxin, Performance, Sterility, Mycoplasma
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.