missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ B-27™ Supplement (50X), serum free
GREENER_CHOICE

Brand:  Gibco™ 17504001

823.88 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 15360284

  • £958.00 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

The most cited neuronal cell culture supplement, B-27 Supplement is an optimized serum-free supplement used to support the low- or high-density growth and short- or long-term viability of embryonic, post-natal, and adult hippocampal and other CNS neurons. B-27 Supplement is provided as a 50X liquid and is intended for use with Neurobasal Medium or Neurobasal-A Medium for neuronal cell culture without the need for an astrocyte feeder layer.

  • Provided as 50X liquid
  • Used with Neurobasal™ Medium for neuronal cell culture
  • Tested in growth assay utilizing rat (Sprague Dawley) embryonic hippocampal neurons
  • Tested also for endotoxin at a 1X concentration
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in freezer (-5 to -20°C) and protect from light.
Serum-free
Mammalian Cell Culture
Cell Culture Supplement
Sterile
100 mL
Neural Cells
10 X
Liquid
Serum-Free
Fewer resources used, Less waste
Endotoxin, Performance, Sterility, Mycoplasma
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.