Learn More
Brady™ B-423 Polyester Thermal Transfer Labels
Printable, polyester labels for Thermal Transfer Printers.
£40.75 - £324.00
Specifications
Autoclavable | Autoclavable |
---|---|
Certifications/Compliance | UL, CSA and AGA Approved |
Adhesive Type | Permanent Acrylic |
Color | White |
Finish | Gloss |
Product Code | Brand | Height (Metric) | Labels per Row | Vertical Repeat (Metric) | Width (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Labels per Row | Vertical Repeat (Metric) | Width (Metric) | Price | Quantity & Availability | ||
11593764
|
Brady™
030191 |
3.2 mm | 2 | 6.4 mm | 38.1 mm |
£131.00
1 roll |
In Stock
Log in to see stock. |
||
11558254
|
Brady™
030507 |
4.8 mm | 1 | 7.3 mm | 25.4 mm |
£131.00
1 roll |
In Stock
Log in to see stock. |
||
11589053
|
Brady™
018654 |
5.1 mm | 1 | 7.6 mm | 12.7 mm |
£80.90
1 roll |
In Stock
Log in to see stock. |
||
11508264
|
Brady™
030512 |
5.1 mm | 1 | 7.6 mm | 16.5 mm |
£81.45
1 roll |
In Stock
Log in to see stock. |
||
11583704
|
Brady™
030120 |
5.1 mm | 4 | 7.6 mm | 16.5 mm |
£81.45
1 roll |
In Stock
Log in to see stock. |
||
11598254
|
Brady™
030511 |
6.4 mm | 1 | 8.9 mm | 19.1 mm |
£118.00
1 roll |
Estimated Shipment: 20-12-2024
Log in to see stock. |
||
11528264
|
Brady™
030514 |
6.4 mm | 1 | 8.9 mm | 22.9 mm |
£111.00
1 roll |
In Stock
Log in to see stock. |
||
11599053
|
Brady™
018658 |
6.4 mm | 1 | 8.9 mm | 25.4 mm |
£129.00
1 roll |
In Stock
Log in to see stock. |
||
11568254
|
Brady™
030508 |
6.4 mm | 1 | 8.9 mm | 31.8 mm |
£125.00
1 roll |
Estimated Shipment: 20-12-2024
Log in to see stock. |
||
11578254
|
Brady™
030509 |
6.4 mm | 1 | 8.9 mm | 34.9 mm |
£137.00
1 roll |
In Stock
Log in to see stock. |
||
11518264
|
Brady™
030513 |
6.4 mm | 1 | 8.9 mm | 50.8 mm |
£182.00
1 roll |
Estimated Shipment: 20-12-2024
Log in to see stock. |
||
11543714
|
Brady™
030126 |
6.4 mm | 2 | 9.5 mm | 31.8 mm |
£125.00
1 roll |
In Stock
Log in to see stock. |
||
11563784
|
Brady™
030218 |
6.4 mm | 2 | 9.5 mm | 34.9 mm |
£137.00
1 roll |
In Stock
Log in to see stock. |
||
11573704
|
Brady™
030119 |
6.4 mm | 3 | 8.9 mm | 22.9 mm |
£111.00
1 roll |
In Stock
Log in to see stock. |
||
11513714
|
Brady™
030123 |
6.4 mm | 12 | 9.5 mm | 6.4 mm |
£40.75
1 roll |
In Stock
Log in to see stock. |
||
11523804
|
Brady™
030237 |
9.5 mm | 2 | 12.1 mm | 31.8 mm |
£150.00
1 roll |
In Stock
Log in to see stock. |
||
15751427
|
Brady™
030106 |
9.5 mm | 3 | 12.1 mm | 25.4 mm |
£125.00
1 roll |
In Stock
Log in to see stock. |
||
11543814
|
Brady™
030250 |
9.5 mm | 4 | 12.1 mm | 17.8 mm |
£78.05
1 roll |
In Stock
Log in to see stock. |
||
11523784
|
Brady™
030214 |
9.5 mm | 7 | 12.1 mm | 9.5 mm |
£87.85
1 roll |
Estimated Shipment: 20-12-2024
Log in to see stock. |
||
11573794
|
Brady™
030232 |
9.7 mm | 2 | 12.7 mm | 27.9 mm |
£177.00
1 roll |
In Stock
Log in to see stock. |
||
16677865
|
Brady™
030357 |
10.2 mm | 1 | 12.7 mm | 50.8 mm |
£249.00
Each |
In Stock
Log in to see stock. |
||
11523714
|
Brady™
030124 |
11.1 mm | 5 | 13.6 mm | 12.7 mm |
£109.00
1 roll |
In Stock
Log in to see stock. |
||
11543804
|
Brady™
030239 |
12.7 mm | 1 | 15.2 mm | 50.8 mm |
£108.00
1 roll |
Estimated Shipment: 20-12-2024
Log in to see stock. |
||
11553704
|
Brady™
030117 |
19.1 mm | 2 | 22.2 mm | 38.1 mm |
£324.00
1 roll |
In Stock
Log in to see stock. |
||
11583794
|
Brady™
030233 |
19.1 mm | 4 | 22.2 mm | 19.1 mm |
£177.00
1 roll |
In Stock
Log in to see stock. |
||
16667865
|
Brady™
030118 |
31.8 mm | 1 | 34.9 mm | 69.9 mm |
£281.00
Each |
Estimated Shipment: 20-12-2024
Log in to see stock. |
||
Description
- Permanent adhesive
- For smooth surfaces
Specifications
Autoclavable | |
Permanent Acrylic | |
Gloss | |
Thermal Transfer | |
Barcode Labeling, Circuit Board Labeling, Equipment Labeling, General Identification, Inventory Labeling | |
0.076 mm |
UL, CSA and AGA Approved | |
White | |
Polyester (B-423) | |
Non-sterile | |
-70°C to +110°C |