missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ B-7594 Polypropylene Hazardous Substance Labels, 25 mm Core
Click to view available options
Includes:
1 Diamond Layout
2 Diamond Layout
4A Diamond Layout
Unit Size:
1 roll
Description
- Supplied with pre-printed red diamonds
- Ready for you to print the required warning, compliant to the latest regulations
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Certifications/Compliance | Compliant with Directive 2011/65/EU and 2015/863 |
| Adhesive Type | Acrylic, Permanent |
| Coating | None |
| Color | Red on White |
| Finish | Gloss |
| Format | Die-cut |
| Material | Polypropylene (B-7594) |
| Printable Sides | Single-side Printable |
| Print Technology | Thermal Transfer |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction