Learn More
BUCHI FlashPure EcoFlex Cartridges, Silica Solid Loader
FlashPure cartridges are offered in a wide range of sizes, covering different stationary phases, particle sizes and geometries. This enables the user to choose the flash cartridge which best suits his purification needs.
Description
Description
  • Different sizes for scalability from milligrams to grams
  • Broad range of different normal and reversed phases
  • Small and spherical particles for fast separations and high loadings
  • Compatibility with the Pure Navigator software (flash method optimization by interpreting TLC or HPLC scouting runs). 
  • RFID tag for an automatic transfer of important parameters for a run on a Pure chromatography system.
  • Loading capacity: 0.1 - 10% 
  • Endcapped: No
Compliance
UL, CE, CSA
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
BUCHI FlashPure EcoFlex Cartridges, Silica Solid Loader