Learn More
Bollé Safety B-Clean Lens Cleaning Products/Kits
Product Code. 12699090
Click to view available options
Product Type:
Anti-Fog Kit
Anti-Fog Spray
Cleaning Station
Lens Cleaner Spray
Multi-function Dry Cleaning Tissues
Tissue Dispenser
Quantity:
1 Ea.
100/Pk.
200/Pk.
30 mL
50 mL
500 mL
Unit Size:
30mL
500mL
50mL
Each
Pack of 100
Pack of 200
Product Code. 12699090
Brand: Bollé Safety PACW100
Only null left
Add to basket
Add to basket
B-Clean Bollé Safety range of cleaning solutions guarantees optimum cleaning. Alcohol and silicone free, its water-based formula guarantees a perfect cleaning and protection for your spectacles and goggles.
  • Alcohol and silicone free
  • Water-based formula guarantees a perfect cleaning and protection for your spectacles and goggles
TRUSTED_SUSTAINABILITY
Specifications
Product Type Tissue Dispenser
Quantity 100/Pk.