missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B-Myb Antibody (5G5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00004605-M03
This item is not returnable.
View return policy
Description
B-Myb Monoclonal antibody specifically detects B-Myb in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| B-Myb | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| B-MYB, BMYBMGC15600, Myb-like protein 2, myb-related protein B, v-myb avian myeloblastosis viral oncogene homolog-like 2, v-myb myeloblastosis viral oncogene homolog (avian)-like 2 | |
| MYBL2 (AAH53555, 601 a.a. ~ 700 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. TLPKSLSLPTTAPSNSSSLTLSGIKEDNSLLNQGFLQAKPEKAAVAQKPRSHFTTPAPMSSAWKTVACGGTRDQLFMQEKARQLLGRLKPSHTSRTLILS | |
| 0.1 mg | |
| Apoptosis, Cancer, Core ESC Like Genes, Stem Cell Markers | |
| 4605 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Immunocytochemistry | |
| 5G5 | |
| Western Blot 1:500 | |
| AAH53555 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction