missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ B-PER™ Bacterial Protein Extraction Reagent

Efficiently lyse E.coli and other bacterial cells and gently extract native proteins with these nonionic detergent solutions.

Brand:  Thermo Scientific™ 90084

190.00 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.


 View more versions of this product

Product Code. 10733485

  • £285.00 / 250mL
In Stock
to see stock.
Explore more special offers
Add to basket

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

B-PER Bacterial Protein Extraction Reagent
Bacterial Protein Extraction Reagent
Store B-PER Reagent at ambient temperature. Store Lysozyme and DNaseI at -20°C.
250 mL
250 mL
B-PER™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZJ403861 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number90084
Lot NumberZF396291 Certificate TypeCertificate of Analysis Date03/07/2024 Catalog Number90084
Lot NumberZC391563 Certificate TypeCertificate of Analysis Date12/04/2024 Catalog Number90084
Lot NumberZB389431 Certificate TypeCertificate of Analysis Date14/02/2024 Catalog Number90084
Lot NumberYJ381917 Certificate TypeCertificate of Analysis Date21/10/2023 Catalog Number90084
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ B-PER™ Bacterial Protein Extraction Reagent > 250 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.