Learn More
Thermo Scientific™ B-PER™ Bacterial Protein Extraction Reagent
Efficiently lyse E.coli and other bacterial cells and gently extract native proteins with these nonionic detergent solutions.
£216.00 - £465.00
Specifications
Description B-PER Bacterial Protein Extraction Reagent
Product Type Bacterial Protein Extraction Reagent
Product Line B-PER™
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11834091
View Documents
Thermo Scientific™
78243
165 mL
£216.00
165mL
In Stock
to see stock.
Add to basket
10541104
View Documents Promotion Details
Thermo Scientific™
78248
500 mL
£465.00
500mL
In Stock
to see stock.
Edge
Add to basket
10733485
View Documents Promotion Details
Thermo Scientific™
90084
250 mL
£266.00
250mL
In Stock
to see stock.
Edge
Add to basket
Description
Description
Perform efficient, gentle lysis and extraction of E.coli and other bacterial cells with Thermo Scientific™ B-PER Bacterial Protein Extraction Reagents.
These easy-to-use cell lysis reagents are nonionic detergent solutions that effectively disrupt cells and solubilize native or recombinant proteins without denaturation. Certain formulations include DNase I and lysozyme enzyme supplements to improve yield of large molecular weight proteins that are difficult to purify. B-PER Reagents are compatible with the addition of standard protease inhibitors and with downstream protein purification methods, such as GST purification and His-tagged immobilized metal affinity chromatography (IMAC).
Highlights:
  • Ready to use – B-PER Reagents provide one-step E.coli cell lysis by a mild, nonionic detergent (proprietary) in your choice of Tris or phosphate buffer formulations
  • Fast and simple – just add B-PER Reagent to a bacterial pellet, shake for 10 minutes and recover soluble proteins after pelleting the cell debris
  • Excellent yields – recover both soluble and insoluble recombinant protein from bacterial lysates and purify inclusion bodies to near-homogeneous levels
  • Flexible – B-PER Reagents are suitable for any scale of protein extraction and are available in phosphate and 1X and 2X Tris formulations, with and without enzymes
  • Compatible – completely compatible with addition of protease inhibitor cocktails, and resulting protein extract can be used in protein assays, typical affinity purification methods (e.g., GST, 6xHis) and other applications
Convenient, ready-to-use formats:
  • B-PER and B-PER II (2X) Reagents are free of enzymatic components
  • B-PER Reagent (in phosphate buffer) is an amine-free formulation for direct compatibility with labeling and crosslinking
  • B-PER with Enzymes and B-PER Direct with Enzymes include lysozyme and DNase I for improved lysis and recovery
Includes:
Reagent packages are available in Tris and phosphate buffer formulations; Kits contain Tris-based B-PER Reagent plus lysozyme and DNase I enzymes.
Specifications
Specifications
B-PER Bacterial Protein Extraction Reagent
Bacterial Protein Extraction Reagent
B-PER™