Learn More
Alpha Packaging™ B-Safe Biohazard Bags
Use Alpha Packaging™ B-Safe Biohazard Bags for heavy duty biohazardous waste. The bags are yellow with biohazard printed on them.
Brand: Alpha Packaging™ 12310609
33.60 GBP valid until 2024-07-31
Use promo code "21640" to get your promotional price.
Code : 04
Additional Details : Weight : 3.68100kg
Description
Bench top dispenser boxesSpecifications
Waste Disposal Bags | |
Waste Disposal | |
5 mil |
Yellow | |
Polyethylene | |
760 x 455 mm |